siRNA Details
Gene Symbol: Senp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012355.1 | NP_001012355.1 | 3 | 416 | ||
XM_006243196.2 | XP_006243258.1 | 2 | 510 | |||
GenBank mRNA | BC089939.1 | - | 3 | 416 | 7310036 | |
FQ226292.1 | - | 2 | 398 | |||
FQ231841.1 | - | 2 | 408 |
