siRNA Details
| Gene Symbol: Senp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012355.1 | NP_001012355.1 | 3 | 535 | ||
| XM_006243196.2 | XP_006243258.1 | 2 | 629 | |||
| GenBank mRNA | BC089939.1 | - | 3 | 535 | 7310036 | |
| FQ226292.1 | - | 2 | 517 | |||
| FQ227853.1 | - | 3 | 548 | |||
| FQ231841.1 | - | 2 | 527 | |||

Senp8
Genomic Map