siRNA Details
| Gene Symbol: Fem1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108157.1 | NP_001101627.1 | 2 | 1934 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fem1b| Gene Symbol: Fem1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108157.1 | NP_001101627.1 | 2 | 1934 | ||