siRNA Details
| Gene Symbol: LOC103690937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758662.1 | XP_008756884.1 | 2 | 1266 | ||
| Gene Symbol: Fem1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108157.1 | NP_001101627.1 | 2 | 1960 | ||

LOC103690937
Genomic Map