siRNA Details
| Gene Symbol: Fam81a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108163.1 | NP_001101633.1 | 5, 6 | 899 | ||
| XM_006243378.2 | XP_006243440.1 | 5, 6 | 735 | |||
| XM_006243379.2 | XP_006243441.1 | 5, 6 | 807 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam81a| Gene Symbol: Fam81a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108163.1 | NP_001101633.1 | 5, 6 | 899 | ||
| XM_006243378.2 | XP_006243440.1 | 5, 6 | 735 | |||
| XM_006243379.2 | XP_006243441.1 | 5, 6 | 807 | |||