siRNA Details
| Gene Symbol: Zfp280d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108165.2 | NP_001101635.2 | 15, 16 | 2082 | ||
| XM_006243356.2 | XP_006243418.1 | 14, 15 | 2049 | |||
| XM_006243357.2 | XP_006243419.1 | 15, 16 | 2045 | |||
| XM_008766288.1 | XP_008764510.1 | 14, 15 | 2032 | |||
| XM_008766289.1 | XP_008764511.1 | 16, 17 | 2225 | |||
| XM_008766290.1 | XP_008764512.1 | 14, 15 | 2032 | |||
| GenBank mRNA | FQ222707.1 | - | 16, 17 | 2230 | ||

Zfp280d
Genomic Map