siRNA Details
| Gene Symbol: Phip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243470.1 | XP_006243532.1 | 26 | 3271 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phip| Gene Symbol: Phip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243470.1 | XP_006243532.1 | 26 | 3271 | ||