siRNA Details
Gene Symbol: RGD1309079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134472.1 | NP_001127944.1 | 3 | 1267 | ||
GenBank mRNA | AY325189.1 | - | 390 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1309079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134472.1 | NP_001127944.1 | 3 | 1267 | ||
GenBank mRNA | AY325189.1 | - | 390 |