siRNA Details
| Gene Symbol: U2surp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729954.4 | XP_002730000.1 | 16 | 1690 | ||
| XM_006243554.1 | XP_006243616.1 | 17 | 1835 | |||
| XM_236501.7 | XP_236501.3 | 17 | 1876 | |||
| GenBank mRNA | FQ226547.1 | - | 17 | 1844 | ||

U2surp
Genomic Map