siRNA Details
| Gene Symbol: Nrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191797.1 | NP_001178726.1 | 4192 | |||
| XM_008773418.1 | XP_008771640.1 | 3792 | ||||
| XM_008773463.1 | XP_008771685.1 | 2075 | ||||
| XM_008773464.1 | XP_008771686.1 | 10 | 1930 | |||

Nrk
Genomic Map