siRNA Details
| Gene Symbol: Nrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191797.1 | NP_001178726.1 | 1940 | |||
| XM_008773418.1 | XP_008771640.1 | 1396 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrk| Gene Symbol: Nrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191797.1 | NP_001178726.1 | 1940 | |||
| XM_008773418.1 | XP_008771640.1 | 1396 | ||||