siRNA Details
| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773419.1 | XP_008771641.1 | 1 | 562 | ||
| GenBank mRNA | BC160840.1 | - | 1 | 350 | 7460350 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf128| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773419.1 | XP_008771641.1 | 1 | 562 | ||
| GenBank mRNA | BC160840.1 | - | 1 | 350 | 7460350 | |