siRNA Details
| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173349.1 | NP_001166820.1 | 1 | 270 | ||
| GenBank mRNA | BC167061.1 | - | 1 | 270 | 8364382 | |
| FQ210020.1 | - | 1 | 258 | |||
| FQ218340.1 | - | 1 | 258 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf128| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173349.1 | NP_001166820.1 | 1 | 270 | ||
| GenBank mRNA | BC167061.1 | - | 1 | 270 | 8364382 | |
| FQ210020.1 | - | 1 | 258 | |||
| FQ218340.1 | - | 1 | 258 | |||