siRNA Details
| Gene Symbol: Tbc1d8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749480.3 | XP_003749528.1 | 12 | 2186 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d8b| Gene Symbol: Tbc1d8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749480.3 | XP_003749528.1 | 12 | 2186 | ||