siRNA Details
| Gene Symbol: Armc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173354.2 | NP_001166825.1 | 16 | 1516 | ||
| XM_006243622.2 | XP_006243684.1 | 16 | 1753 | |||
| XM_006243623.2 | XP_006243685.1 | 14 | 1588 | |||
| XM_006243624.2 | XP_006243686.1 | 15 | 1552 | |||
| XM_006243625.2 | XP_006243687.1 | 15 | 1592 | |||
| GenBank mRNA | BC168192.1 | - | 16 | 1424 | ||
| FQ211639.1 | - | 15 | 1579 | |||

Armc8
Genomic Map