siRNA Details
| Gene Symbol: Armc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173354.2 | NP_001166825.1 | 13, 14 | 1310 | ||
| XM_006243622.2 | XP_006243684.1 | 13, 14 | 1547 | |||
| XM_006243624.2 | XP_006243686.1 | 12, 13 | 1346 | |||
| XM_006243625.2 | XP_006243687.1 | 12, 13 | 1386 | |||
| GenBank mRNA | BC168192.1 | - | 13, 14 | 1218 | ||
| FQ211639.1 | - | 12, 13 | 1373 | |||

Armc8
Genomic Map