siRNA Details
| Gene Symbol: Acad11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108181.1 | NP_001101651.1 | 5 | 686 | ||
| GenBank mRNA | BC167762.1 | - | 5 | 691 | 7443858 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acad11| Gene Symbol: Acad11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108181.1 | NP_001101651.1 | 5 | 686 | ||
| GenBank mRNA | BC167762.1 | - | 5 | 691 | 7443858 | |