siRNA Details
| Gene Symbol: Dock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750582.3 | XP_003750630.1 | 18 | 2125 | ||
| XM_006243865.2 | XP_006243927.1 | 18 | 2125 | |||
| XM_006243866.2 | XP_006243928.1 | 18 | 2125 | |||
| XM_006243867.2 | XP_006243929.1 | 18 | 2125 | |||
| XM_006243868.2 | XP_006243930.1 | 18 | 2125 | |||

Dock3
Genomic Map