siRNA Details
Gene Symbol: Rbm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108186.1 | NP_001101656.1 | 3 | 838 | ||
XM_006243780.2 | XP_006243842.1 | 4 | 934 | |||
XM_006243781.2 | XP_006243843.1 | 3 | 1107 | |||
XM_008766534.1 | XP_008764756.1 | 3 | 1073 |
