siRNA Details
| Gene Symbol: Rbm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108186.1 | NP_001101656.1 | 20 | 3447 | ||
| XM_006243780.2 | XP_006243842.1 | 21 | 3543 | |||
| XM_006243781.2 | XP_006243843.1 | 20 | 3716 | |||
| XM_008766534.1 | XP_008764756.1 | 20 | 3682 | |||
| XM_008766535.1 | XP_008764757.1 | 15 | 2412 | |||
| GenBank mRNA | FQ229938.1 | - | 8 | 928 | ||

Rbm6
Genomic Map