siRNA Details
| Gene Symbol: Arih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012275.2 | NP_001012275.1 | 13 | 1365 | ||
| XM_006243784.2 | XP_006243846.1 | 13 | 1833 | |||
| XM_006243785.2 | XP_006243847.1 | 13 | 1830 | |||
| XM_006243787.2 | XP_006243849.1 | 12 | 1054 | |||
| XM_008766537.1 | XP_008764759.1 | 13 | 1518 | |||
| XM_008766538.1 | XP_008764760.1 | 13 | 1521 | |||
| GenBank mRNA | CV114328.1 | - | 2 | 233 | ||

Arih2
Genomic Map