siRNA Details
| Gene Symbol: Ccdc51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014098.1 | NP_001014120.1 | 2 | 254 | ||
| XM_006243788.2 | XP_006243850.1 | 2 | 458 | |||
| XM_006243789.2 | XP_006243851.1 | 2 | 1032 | |||
| XM_006243790.2 | XP_006243852.1 | 2 | 532 | |||
| XM_006243791.2 | XP_006243853.1 | 2 | 836 | |||
| XM_006243792.2 | XP_006243854.1 | 2 | 414 | |||
| GenBank mRNA | BC087581.1 | - | 2 | 254 | 7314505 | |

Ccdc51
Genomic Map