siRNA Details
Gene Symbol: Nbeal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750590.3 | XP_003750638.1 | 48 | 7684 | ||
XM_003750591.3 | XP_003750639.1 | 47 | 7603 | |||
XM_006244008.2 | XP_006244070.1 | 48 | 8911 | |||
XM_006244009.2 | XP_006244071.1 | 49 | 7610 |
