siRNA Details
| Gene Symbol: Dclk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191800.1 | NP_001178729.1 | 3 | 2035 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dclk3| Gene Symbol: Dclk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191800.1 | NP_001178729.1 | 3 | 2035 | ||