siRNA Details
| Gene Symbol: Trak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134565.1 | NP_001128037.1 | 12 | 1201 | ||
| XM_006244099.2 | XP_006244161.1 | 11 | 2282 | |||
| XM_006244100.2 | XP_006244162.1 | 12 | 1208 | |||
| XM_006244101.2 | XP_006244163.1 | 12 | 3992 | |||
| XM_006244102.2 | XP_006244164.1 | 11 | 2629 | |||
| XM_006244103.2 | XP_006244165.1 | 13 | 1624 | |||
| XM_008766679.1 | XP_008764901.1 | 11 | 2311 | |||
| XM_008766680.1 | XP_008764902.1 | 11 | 1695 | |||

Trak1
Genomic Map