siRNA Details
| Gene Symbol: Abhd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212524.1 | NP_997689.1 | 5, 6 | 824 | ||
| XM_006244107.1 | XP_006244169.1 | 6, 7 | 929 | |||
| XM_006244108.2 | XP_006244170.1 | 5, 6 | 829 | |||
| XM_006244109.2 | XP_006244171.1 | 4, 5 | 2217 | |||
| GenBank mRNA | AY550934.1 | - | 5, 6 | 824 | ||
| BC127470.1 | - | 5, 6 | 858 | |||

Abhd5
Genomic Map