siRNA Details
| Gene Symbol: Gpr108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199399.1 | NP_955431.1 | 2 | 155 | ||
| XM_006244285.2 | XP_006244347.1 | 2 | 205 | |||
| XM_006244286.2 | XP_006244348.1 | 2 | 132 | |||
| GenBank mRNA | BC061996.1 | - | 2 | 155 | 5599699 | |
More Information
Other Species Targeted
Set Membership: |
|

Gpr108
Genomic Map