siRNA Details
| Gene Symbol: Gpr108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199399.1 | NP_955431.1 | 5, 6 | 523 | ||
| GenBank mRNA | BC061996.1 | - | 5, 6 | 523 | 5599699 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr108| Gene Symbol: Gpr108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199399.1 | NP_955431.1 | 5, 6 | 523 | ||
| GenBank mRNA | BC061996.1 | - | 5, 6 | 523 | 5599699 | |