siRNA Details
| Gene Symbol: Taf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108197.1 | NP_001101667.1 | 7 | 773 | ||
| XM_008766860.1 | XP_008765082.1 | 7 | 774 | |||
| XM_008766861.1 | XP_008765083.1 | 7 | 774 | |||
| XM_008766862.1 | XP_008765084.1 | 7 | 774 | |||
| GenBank mRNA | FQ211123.1 | - | 7 | 770 | ||

Taf8
Genomic Map