siRNA Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191582.1 | NP_001178511.1 | 29 | 5607 | ||
| XM_006244518.2 | XP_006244580.1 | 29 | 5931 | |||
| XM_008766864.1 | XP_008765086.1 | 29 | 5931 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cul9| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191582.1 | NP_001178511.1 | 29 | 5607 | ||
| XM_006244518.2 | XP_006244580.1 | 29 | 5931 | |||
| XM_008766864.1 | XP_008765086.1 | 29 | 5931 | |||