siRNA Details
| Gene Symbol: Polh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108204.1 | NP_001101674.1 | 10 | 1403 | ||
| XM_006244537.1 | XP_006244599.1 | 8 | 831 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Polh| Gene Symbol: Polh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108204.1 | NP_001101674.1 | 10 | 1403 | ||
| XM_006244537.1 | XP_006244599.1 | 8 | 831 | |||