siRNA Details
| Gene Symbol: Mad2l1bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009699.1 | NP_001009699.1 | 3 | 810 | ||
| GenBank mRNA | BC088268.1 | - | 3 | 810 | 7379293 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mad2l1bp| Gene Symbol: Mad2l1bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009699.1 | NP_001009699.1 | 3 | 810 | ||
| GenBank mRNA | BC088268.1 | - | 3 | 810 | 7379293 | |