siRNA Details
| Gene Symbol: Tcte1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108206.1 | NP_001101676.1 | 4 | 1464 | ||
| XM_006244539.2 | XP_006244601.1 | 5 | 1987 | |||
| XM_006244540.2 | XP_006244602.1 | 6 | 2064 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcte1| Gene Symbol: Tcte1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108206.1 | NP_001101676.1 | 4 | 1464 | ||
| XM_006244539.2 | XP_006244601.1 | 5 | 1987 | |||
| XM_006244540.2 | XP_006244602.1 | 6 | 2064 | |||