siRNA Details
| Gene Symbol: Enpp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012744.1 | NP_001012762.1 | 3 | 1006 | ||
| XM_006244607.2 | XP_006244669.1 | 3 | 1030 | |||
| GenBank mRNA | BC090331.1 | - | 3 | 1006 | 7300780 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Enpp5| Gene Symbol: Enpp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012744.1 | NP_001012762.1 | 3 | 1006 | ||
| XM_006244607.2 | XP_006244669.1 | 3 | 1030 | |||
| GenBank mRNA | BC090331.1 | - | 3 | 1006 | 7300780 | |