siRNA Details
| Gene Symbol: Cd2ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181475.2 | NP_852140.1 | 15 | 1787 | ||
| GenBank mRNA | AY205155.1 | - | 15 | 1787 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd2ap| Gene Symbol: Cd2ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181475.2 | NP_852140.1 | 15 | 1787 | ||
| GenBank mRNA | AY205155.1 | - | 15 | 1787 | ||