siRNA Details
| Gene Symbol: Pgk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012130.1 | NP_001012130.1 | 1 | 680 | ||
| GenBank mRNA | BC083568.1 | - | 1 | 680 | 7111073 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgk2| Gene Symbol: Pgk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012130.1 | NP_001012130.1 | 1 | 680 | ||
| GenBank mRNA | BC083568.1 | - | 1 | 680 | 7111073 | |