siRNA Details
Gene Symbol: Zfp451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033705.1 | NP_001028877.1 | 10 | 1399 | ||
GenBank mRNA | BC101930.1 | - | 10 | 1399 | 7938755 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033705.1 | NP_001028877.1 | 10 | 1399 | ||
GenBank mRNA | BC101930.1 | - | 10 | 1399 | 7938755 |