siRNA Details
| Gene Symbol: Arid5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034934.1 | NP_001030106.1 | 3 | 126 | ||
| XM_006244806.2 | XP_006244868.1 | 5 | 428 | |||
| XM_006244807.2 | XP_006244869.1 | 5 | 425 | |||
| GenBank mRNA | BC105777.1 | - | 3 | 126 | 7931985 | |

Arid5a
Genomic Map