siRNA Details
| Gene Symbol: Kansl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034835.1 | NP_001030007.1 | 1 | 271 | ||
| XM_006244808.2 | XP_006244870.1 | 2 | 332 | |||
| XM_006244809.2 | XP_006244871.1 | 2 | 385 | |||
| XM_006244810.2 | XP_006244872.1 | 2 | 563 | |||
| XM_008767027.1 | XP_008765249.1 | 2 | 327 | |||
| GenBank mRNA | BC105865.1 | - | 1 | 271 | 7461395 | |

Kansl3
Genomic Map