siRNA Details
| Gene Symbol: Kansl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034835.1 | NP_001030007.1 | 14, 15 | 2075 | ||
| XM_006244808.2 | XP_006244870.1 | 16, 17 | 2214 | |||
| XM_006244809.2 | XP_006244871.1 | 16, 17 | 2267 | |||
| XM_006244810.2 | XP_006244872.1 | 16, 17 | 2445 | |||
| XM_006244811.1 | XP_006244873.1 | 16, 17 | 2181 | |||
| XM_006244812.1 | XP_006244874.1 | 15, 16 | 1928 | |||
| XM_006244813.1 | XP_006244875.1 | 15, 16 | 1833 | |||
| XM_008767027.1 | XP_008765249.1 | 16, 17 | 2209 | |||
| XM_008767028.1 | XP_008765250.1 | 14, 15 | 2056 | |||
| GenBank mRNA | BC105865.1 | - | 14, 15 | 2075 | 7461395 | |

Kansl3
Genomic Map