siRNA Details
| Gene Symbol: Tmem131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244855.2 | XP_006244917.1 | 16 | 1710 | ||
| XM_006244856.2 | XP_006244918.1 | 16 | 1791 | |||
| XM_006244858.2 | XP_006244920.1 | 15 | 3628 | |||
| XM_008767047.1 | XP_008765269.1 | 16 | 1710 | |||
| XM_237056.8 | XP_237056.4 | 16 | 1710 | |||

Tmem131
Genomic Map