siRNA Details
| Gene Symbol: Rev1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108213.1 | NP_001101683.1 | 5 | 707 | ||
| XM_006244819.2 | XP_006244881.1 | 6 | 1439 | |||
| XM_006244820.2 | XP_006244882.1 | 6 | 2059 | |||
| XM_006244821.2 | XP_006244883.1 | 5 | 1241 | |||
| XM_008767030.1 | XP_008765252.1 | 2 | 246 | |||

Rev1
Genomic Map