siRNA Details
| Gene Symbol: Rev1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108213.1 | NP_001101683.1 | 16 | 2744 | ||
| XM_006244819.2 | XP_006244881.1 | 17 | 3476 | |||
| XM_006244820.2 | XP_006244882.1 | 17 | 4096 | |||
| XM_006244821.2 | XP_006244883.1 | 16 | 3275 | |||
| XM_008767030.1 | XP_008765252.1 | 13 | 2283 | |||

Rev1
Genomic Map