siRNA Details
| Gene Symbol: LOC102548228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244873.1 | XP_006244935.1 | 2 | 781 | ||
| XM_006244874.2 | XP_006244936.1 | 2 | 354 | |||
| XM_006244875.2 | XP_006244937.1 | 2 | 569 | |||
| XM_006244876.1 | XP_006244938.1 | 2 | 501 | |||
| XM_006244879.1 | XP_006244941.1 | 2 | 689 | |||
| XM_006244881.2 | XP_006244943.1 | 2 | 478 | |||
| XM_008767062.1 | XP_008765284.1 | 2 | 307 | |||

LOC102548228
Genomic Map