siRNA Details
Gene Symbol: Ccdc150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191806.1 | NP_001178735.1 | 1 | 120 | ||
XM_006244924.2 | XP_006244986.1 | 2 | 342 | |||
XM_008767094.1 | XP_008765316.1 | 2 | 342 | |||
XM_008767095.1 | XP_008765317.1 | 2 | 342 | |||
XM_008767096.1 | XP_008765318.1 | 2 | 342 | |||
XM_008767097.1 | XP_008765319.1 | 2 | 342 |
