siRNA Details
| Gene Symbol: Pgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201990.1 | NP_973719.1 | 8, 9 | 1064 | ||
| GenBank mRNA | AB116149.1 | - | 8, 9 | 1064 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgap1| Gene Symbol: Pgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201990.1 | NP_973719.1 | 8, 9 | 1064 | ||
| GenBank mRNA | AB116149.1 | - | 8, 9 | 1064 | ||