siRNA Details
| Gene Symbol: Aox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008522.1 | NP_001008522.1 | 25 | 2745 | ||
| XM_008767106.1 | XP_008765328.1 | 26 | 4374 | |||
| GenBank mRNA | AY665588.1 | - | 25 | 2745 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aox2| Gene Symbol: Aox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008522.1 | NP_001008522.1 | 25 | 2745 | ||
| XM_008767106.1 | XP_008765328.1 | 26 | 4374 | |||
| GenBank mRNA | AY665588.1 | - | 25 | 2745 | ||