siRNA Details
Gene Symbol: Ica1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199400.1 | NP_955432.1 | 2, 3 | 229 | ||
XM_006244994.2 | XP_006245056.1 | 3, 4 | 376 | |||
XM_006244995.2 | XP_006245057.1 | 3, 4 | 376 | |||
XM_006244998.2 | XP_006245060.1 | 3, 4 | 376 | |||
XM_006244999.2 | XP_006245061.1 | 3, 4 | 376 | |||
XM_008767111.1 | XP_008765333.1 | 3, 4 | 358 | |||
XM_008767112.1 | XP_008765334.1 | 3, 4 | 376 | |||
GenBank mRNA | AY491990.1 | - | 2, 3 | 229 |
