siRNA Details
| Gene Symbol: Smarcal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108222.1 | NP_001101692.1 | 4 | 1130 | ||
| XM_006245212.2 | XP_006245274.1 | 5 | 1542 | |||
| XM_006245213.2 | XP_006245275.1 | 5 | 1066 | |||
| XM_008767235.1 | XP_008765457.1 | 5 | 1206 | |||
| XM_008767236.1 | XP_008765458.1 | 5 | 1430 | |||
| XM_008767237.1 | XP_008765459.1 | 5 | 576 | |||
| GenBank mRNA | BC168154.1 | - | 5 | 1144 | 8362162 | |

Smarcal1
Genomic Map