siRNA Details
| Gene Symbol: Chpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005906.1 | NP_001005906.1 | 2 | 765 | ||
| XM_006245228.1 | XP_006245290.1 | 2 | 394 | |||
| GenBank mRNA | BC083617.1 | - | 2 | 765 | 7133858 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chpf| Gene Symbol: Chpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005906.1 | NP_001005906.1 | 2 | 765 | ||
| XM_006245228.1 | XP_006245290.1 | 2 | 394 | |||
| GenBank mRNA | BC083617.1 | - | 2 | 765 | 7133858 | |